Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-3-nitrotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Nitrobenzyl chlorideα-Chloro-m-nitrotoluene1-(Chloromethyl)-3-nitrobenzenem-Nitrobenzyl chloridealpha-Chloro-3-nitrotoluene

A16775-18 A16775-30 A16775-09 A16775-30 A16775-18 A16775-09
AAAA16775-30EA 441.99 USD InStock
CAAAA16775-18 CAAAA16775-09 AAAA16775-30 AAA16775-09 AAA16775-18 CAAAAA16775-30
α-Chloro-3-nitrotoluene 98%
α-Chloro-3-nitrotoluene


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 150 °C/3 mm
Melting Pt: 43…47 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007272
CAS Number: 619-23-8
EINECS: 210-586-1
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty