Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Gluconic acid 50% in aqueous solution

Supplier: Thermo Scientific Chemicals
Synonyms: Gluconic acid(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

A16806-AP A16806-0F A16806-0F A16806-AP
AAA16806-0FEA 82 USD InStock
CAAAA16806-AP AAA16806-0F CAAAA16806-0F AAA16806-AP
D-Gluconic acid 50% in aqueous solution
D-Gluconic acid


Formula: HOH₂C(CH(OH))₄COOH
MW: 196.16 g/mol
Density: 1.234
Storage Temperature: Ambient
MDL Number: MFCD00004240
CAS Number: 526-95-4
EINECS: 208-401-4
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty