Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-D-Allose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: β-D-Allopyranose(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol

A16810-MC A16810-ME A16810-ME A16810-MC
AAA16810-MCEA 56.89 USD InStock
CAAAA16810-ME CAAAA16810-MC AAA16810-MC AAA16810-ME
β-D-Allose 98%
β-D-Allose


Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00063268
CAS Number: 7283-09-2
EINECS: 219-994-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty