Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Allopurinol

A16974-06 A16974-14 A16974-06 A16974-14
AAA16974-14EA 136.99 USD InStock
AAA16974-14 AAA16974-06 CAAAA16974-14 CAAAA16974-06
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
Xanthine oxidase inhibitor; decreases uric acid production


Formula: C5H4N4O
MW: 136.11 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00599413
CAS Number: 315-30-0
EINECS: 206-250-9
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty