Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Lyxose 99%

Supplier: Thermo Scientific Chemicals

A17007-14 A17007-14 A17007-06 A17007-03 A17007-06 A17007-03
AAA17007-06EA 71.39 USD InStock
CAAAA17007-06 CAAAA17007-14 CAAAA17007-03 AAA17007-06 AAA17007-14 AAA17007-03
D-(-)-Lyxose 99%
D(-)-Lyxose


Formula: C₅H₁₀O₅
MW: 150.13
Melting Pt: 109…115 °C
Density: 1.545
Storage Temperature: Ambient
MDL Number: MFCD00064362
CAS Number: 1114-34-7
EINECS: 214-212-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty