Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylimidazole 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methyl-1H-imidazole

A17063-36 A17063-0E A17063-0E A17063-36 A17063-22 A17063-22
AAA17063-22EA 47.21 USD InStock
AAA17063-22 CAAAA17063-0E CAAAA17063-36 AAA17063-36 AAA17063-0E CAAAA17063-22
2-Methylimidazole 97%
2-Methylimidazole


Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 142…144 °C
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3263
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty