Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Indoxyl sulfate potassium salt 97%

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Indolyl sulphate potassium salt

A17079-06 A17079-03 A17079-01 A17079-01 A17079-06 A17079-03
AAA17079-01EA 70.29 USD InStock
CAAAA17079-03 AAA17079-01 CAAAA17079-01 AAA17079-03 AAA17079-06 CAAAA17079-06
3-Indoxyl sulfate potassium salt 97%
3-Indoxyl sulfate potassium salt


Formula: C₈H₆KNO₄S
MW: 251.31 g/mol
Melting Pt: 165 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00037931
CAS Number: 2642-37-7
EINECS: 220-145-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty