Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triacetin (glycerol triacetate) 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Glycerol triacetate Glyceryl triacetate

A17132-36 A17132-36 A17132-0E A17132-0E
AAA17132-0EEA 169.68 USD InStock
AAA17132-0E CAAAA17132-36 CAAAA17132-0E AAA17132-36
Triacetin (glycerol triacetate) 99%
Triacetin (Glycerol triacetate)


Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 257…259 °C
Melting Pt: 2…4 °C
Density: 1.155
Flash Pt: 149 °C(300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008716
CAS Number: 102-76-1
EINECS: 203-051-9
Merck Index: 12,09721
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty