Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-aminopropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-β-Ala-OEt·HCl3-Aminopropionic Acid Ethyl Ester HydrochlorideEthyl 3-Aminopropionate Hydrochloride

A17277-18 A17277-09 A17277-09 A17277-18
AAA17277-09EA 61.39 USD InStock
AAA17277-09 AAA17277-18 CAAAA17277-18 CAAAA17277-09
Ethyl-3-aminopropanoate hydrochloride 98%
Ethyl-3-aminopropanoate hydrochloride


Formula: C5H12ClNO2
MW: 153.6 g/mol
Melting Pt: 67…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00012909
CAS Number: 4244-84-2
EINECS: 224-203-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty