Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-tyrosine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ac-Tyr-OH

A17307-18 A17307-09 A17307-30 A17307-30 A17307-09 A17307-18
AAA17307-30EA 728.99 USD InStock
CAAAA17307-18 CAAAA17307-09 CAAAA17307-30 AAA17307-30 AAA17307-18 AAA17307-09
N-Acetyl-L-tyrosine 99%
N-Acetyl-L-tyrosine


Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 150…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00037190
CAS Number: 537-55-3
EINECS: 208-671-3
Order Now
Specification Test Results
Optical rotation +58° (c=5 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty