Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylhexylamine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N,N-Dimethylhexan-1-amine

A17364-14 A17364-06 A17364-14 A17364-22 A17364-06 A17364-22
AAA17364-22EA 592.99 USD InStock
CAAAA17364-22 AAA17364-22 AAA17364-14 AAA17364-06 CAAAA17364-14 CAAAA17364-06
N,N-Dimethylhexylamine 99%
N,N-Dimethylhexylamine


Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 146…150 °C
Density: 0.744
Flash Pt: 33 °C(92 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009522
CAS Number: 4385-04-0
EINECS: 224-491-8
UN: 2734
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty