Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyl-1-phenylhydrazine hydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-Benzyl-N-phenylhydrazine hydrochloride

A17367-06 A17367-14 A17367-14 A17367-06
AAA17367-14EA 176.99 USD InStock
AAA17367-14 CAAAA17367-14 AAA17367-06 CAAAA17367-06
1-Benzyl-1-phenylhydrazine hydrochloride 98+%
1-Benzyl-1-phenylhydrazine hydrochloride


Formula: C₁₃H₁₅ClN₂
Melting Pt: 178…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00050690
CAS Number: 5705-15-7
EINECS: 227-195-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty