Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA tetrasodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Edetic acid tetrasodium salt hydrate2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate hydrateEthylenedinitrilotetra-acetic acid tetrasodium salt hydrateTetrasodium ethylenediaminetetraacetate hydrateKestranal® 4S hydrate

A17385-0I A17385-30 A17385-0I A17385-30 A17385-0B A17385-0B
AAA17385-0IEA 241.42 USD InStock
AAA17385-0I CAAAA17385-30 AAA17385-0B CAAAA17385-0B CAAAA17385-0I AAA17385-30
EDTA tetrasodium salt hydrate 98%
EDTA tetrasodium salt hydrate


Formula: C₁₀H₁₄N₂Na₄O₉
MW: 380.17 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150025
CAS Number: 194491-31-1
EINECS: 200-573-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty