Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloromethyl chloroformate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chloroformic acid trichloromethyl ester Diphosgene

A17444-09 A17444-30 A17444-18 A17444-09 A17444-30 A17444-18
AAA17444-09EA 62.49 USD InStock
AAA17444-09 CAAAA17444-30 CAAAA17444-09 AAA17444-30 CAAAA17444-18 AAA17444-18
Trichloromethyl chloroformate 98%
Trichloromethyl chloroformate


Formula: C₂Cl₄O₂
MW: 197.83 g/mol
Boiling Pt: 128 °C
Melting Pt: –57 °C
Density: 1.640
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015553
CAS Number: 503-38-8
EINECS: 207-965-9
UN: 3390
ADR: "6.1",I
Merck Index: 13,03368
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty