Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethyl-m-toluamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Deet Diethyl-m-toluamide N,N-Diethyl-3-methylbenzamide Diethyltoluamide

A17486-0E A17486-22 A17486-36 A17486-36 A17486-22 A17486-0E
AAA17486-22EA 33.44 USD InStock
CAAAA17486-36 CAAAA17486-22 AAA17486-22 AAA17486-36 AAA17486-0E CAAAA17486-0E
N,N-Diethyl-m-toluamide 97%
N,N-Diethyl-m-toluamide


Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 110…112 °C/1 mm
Melting Pt: –45 °C
Density: 0,997
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty