Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol green sodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: Bromocresol green water soluble

A17503-14 A17503-22 A17503-06 A17503-06 A17503-14 A17503-22
AAAA17503-22EA 495.93 USD InStock
AAAA17503-22 AAA17503-06 CAAAAA17503-22 CAAAA17503-14 AAA17503-14 CAAAA17503-06
Bromocresol green sodium salt
Bromocresol green sodium salt


Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty