Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Hydratropic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-2-Phenylpropanoic acid(R)-(-)-2-Phenylpropionic acid(R)-(-)-2-Phenylpropanoic acid

A17639-01 A17639-06 A17639-03 A17639-01 A17639-06 A17639-03
AAAA17639-06EA 626.04 USD InStock
CAAAA17639-03 CAAAA17639-01 CAAAAA17639-06 AAAA17639-06 AAA17639-03 AAA17639-01
(R)-(-)-Hydratropic acid 97%
(R)-(-)-Hydratropic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264…266 °C
Melting Pt: 32…34 °C
Density: 1,100
Flash Pt: 148 °C(298 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063140
CAS Number: 7782-26-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty