Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Imidazolidinone hemihydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Imidazolidone hemihydrate

A17666-0B A17666-30 A17666-0B A17666-30
AAA17666-30EA 46.59 USD InStock
AAA17666-30 CAAAA17666-0B CAAAA17666-30 AAA17666-0B
2-Imidazolidinone hemihydrate 98+%
2-Imidazolidinone hemihydrate


Formula: C₃H₆N₂O·0,5H₂O
MW: 95.10 (86.09 anhy)
Melting Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD02662355
CAS Number: 121325-67-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty