Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stearic acid 90+%

Supplier: Thermo Scientific Chemicals
Synonyms: Octadecanoic acid Octadecylic acid

A17673-0I A17673-0B A17673-0I A17673-0B
AAA17673-0IEA 128.69 USD InStock
AAA17673-0I CAAAA17673-0B AAA17673-0B CAAAA17673-0I
Stearic acid 90+%
Stearic acid


Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 232 °C/15 mm
Melting Pt: 67…70 °C
Density: 0.941
Flash Pt: 196 °C(384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://us-legacy.vwr.com/stibo/web/std.lang.all/40/06/724006.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty