Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heptanoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Heptanoic acid Heptylic acid

A17704-0F A17704-AP A17704-AP A17704-0F
AAA17704-0FEA 240.99 USD InStock
AAA17704-0F CAAAA17704-0F AAA17704-AP CAAAA17704-AP
Heptanoic acid 98+%
Heptanoic acid


Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 222…223 °C
Melting Pt: –8 °C
Density: 0.917
Flash Pt: 114 °C(237 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004426
CAS Number: 111-14-8
EINECS: 203-838-7
UN: 3265
ADR: "8",III
Merck Index: 13,04680
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty