Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Tyrosine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-D-Tyr-OH

A17709-14 A17709-06 A17709-06 A17709-03 A17709-03 A17709-14
AAA17709-03EA 33.7 USD InStock
AAA17709-03 CAAAAA17709-14 AAAA17709-14 CAAAA17709-06 AAA17709-06 CAAAA17709-03
D-(+)-Tyrosine 99%
D(+)-Tyrosine


Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 310…314 °C
Storage Temperature: Ambient
MDL Number: MFCD00063073
CAS Number: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty