Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pyrenebutyric acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A17760-03 A17760-06 A17760-06 A17760-03
AAA17760-03EA 50.29 USD InStock
AAA17760-03 AAA17760-06 CAAAA17760-06 CAAAA17760-03
1-Pyrenebutyric acid 97%
1-Pyrenebutyric acid


Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00004141
CAS Number: 3443-45-6
EINECS: 222-354-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty