Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A17762-14 A17762-22 A17762-06 A17762-22 A17762-06 A17762-14
AAA17762-06EA 37.39 USD InStock
CAAAA17762-06 CAAAAA17762-22 AAA17762-06 AAA17762-14 AAAA17762-22 CAAAA17762-14
2-Chlorobenzothiazole 98%
2-Chlorobenzothiazole


Formula: C₇H₄ClNS
MW: 169.63 g/mol
Boiling Pt: 141 °C/30 mm
Melting Pt: 30…33 °C
Density: 1.370
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005776
CAS Number: 615-20-3
EINECS: 210-415-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty