Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrochalcone 97%

Supplier: Thermo Scientific Chemicals
Warning

A17763-06 A17763-06 A17763-14 A17763-14
AAA17763-14EA 241.99 USD InStock
CAAAA17763-06 AAA17763-14 AAA17763-06 CAAAA17763-14
2-Nitrochalcone 97%
2-Nitrochalcone


Formula: C₁₅H₁₁NO₃
MW: 253.26 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00031069
CAS Number: 7473-93-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty