Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Arabitol 99%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Arabitol(2R,4R)-Pentane-1,2,3,4,5-pentaol

A17801-14 A17801-14 A17801-06 A17801-06
AAA17801-06EA 52.99 USD InStock
AAA17801-06 CAAAA17801-14 CAAAA17801-06 AAA17801-14
D-(+)-Arabitol 99%
D(+)-Arabitol


Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004709
CAS Number: 488-82-4
EINECS: 207-686-2
Order Now
Specification Test Results
Optical rotation +11° (c=5 in 8% borax solution)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty