Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Isoleucine methyl ester hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Ile-OMe.HCl

A17847-14 A17847-06 A17847-14 A17847-03 A17847-03 A17847-06
AAA17847-14EA 515.99 USD InStock
AAA17847-14 AAA17847-03 CAAAA17847-06 AAA17847-06 CAAAA17847-14 CAAAA17847-03
L-Isoleucine methyl ester hydrochloride ≥98%
L-Isoleucine methyl ester hydrochloride


Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00038911
CAS Number: 18598-74-8
EINECS: 242-437-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty