Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-chloroaniline 98+%

Supplier: Thermo Scientific Chemicals
Warning

A17859-18 A17859-30 A17859-18 A17859-30 A17859-09 A17859-09
AAA17859-18EA 150.99 USD InStock
CAAAA17859-09 CAAAA17859-18 AAA17859-18 AAAA17859-30 CAAAAA17859-30 AAA17859-09
4-Bromo-2-chloroaniline 98+%
4-Bromo-2-chloroaniline


Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00007660
CAS Number: 38762-41-3
EINECS: 254-118-4
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty