Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Malic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Hydroxy-1,4-butanedioic acid (±)-Hydroxysuccinic acid

A17874-36 A17874-36 A17874-0E A17874-0E
AAA17874-0EEA 97.59 USD InStock
AAA17874-0E AAA17874-36 CAAAA17874-36 CAAAA17874-0E
DL-Malic acid 98%
DL-Malic acid


Formula: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Melting Pt: 130…133 °C
Density: 1.609
Flash Pt: 203 °C(397 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 230-022-8
Merck Index: 13,05730
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/67/2343067.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty