Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Ribose 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R,4R)-2,3,4,5-Tetrahydroxypentanal

A17894-18 A17894-30 A17894-09 A17894-30 A17894-18 A17894-09
AAA17894-09EA 29.3 USD InStock
CAAAA17894-18 CAAAA17894-09 CAAAA17894-30 AAA17894-09 AAA17894-18 AAA17894-30
D-(-)-Ribose 98%
D(-)-Ribose


Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 82…86 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty