Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Triethylamine hydrobromide

A17926-36 A17926-22 A17926-22 A17926-36
AAA17926-22EA 44.69 USD InStock
AAA17926-22 CAAAA17926-22 CAAAA17926-36 AAA17926-36
Triethylammonium bromide 98%
Triethylammonium bromide
Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.


Formula: C₆H₁₅N.HBr
MW: 182.1 g/mol
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00054287
CAS Number: 636-70-4
EINECS: 211-263-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty