Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Cycloserine 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-(+)-Cycloserine D-4-Aminoisoxazolidin-3-one

A18000-03 A18000-06 A18000-03 A18000-06
AAA18000-03EA 114.23 USD InStock
AAA18000-03 AAA18000-06 CAAAA18000-06 CAAAA18000-03
D-Cycloserine 98+%
D-Cycloserine
Inhibits bacterial cell wall biosynthesis. Prevents conversion of D-Alanine to L-Alanine


Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Melting Pt: 137 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005353
CAS Number: 68-41-7
EINECS: 200-688-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty