Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinobenzothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydrazinylbenzo[d]thiazole

A18006-18 A18006-09 A18006-18 A18006-09
AAA18006-18EA 452.99 USD InStock
CAAAA18006-18 AAA18006-18 CAAAA18006-09 AAA18006-09
2-Hydrazinobenzothiazole 97%
2-Hydrazinobenzothiazole


Formula: C₇H₇N₃S
MW: 165.21 g/mol
Melting Pt: 198…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00041849
CAS Number: 615-21-4
EINECS: 210-416-6
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty