Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-3-methoxybenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A18056-14 A18056-22 A18056-06 A18056-22 A18056-14 A18056-06
AAA18056-06EA 23.9 USD InStock
AAA18056-06 CAAAA18056-22 CAAAA18056-14 CAAAA18056-06 AAA18056-14 AAA18056-22
4-Hydroxy-3-methoxybenzonitrile 98%
4-Hydroxy-3-methoxybenzonitrile


Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001820
CAS Number: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty