Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxybenzophenone 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: m-Hydroxybenzophenone

A18061-03 A18061-06 A18061-03 A18061-14 A18061-14 A18061-06
AAA18061-06EA 177.99 USD InStock
CAAAA18061-03 CAAAA18061-14 AAA18061-06 AAA18061-03 AAA18061-14 CAAAA18061-06
3-Hydroxybenzophenone 99%
3-Hydroxybenzophenone


Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 148 °C (4 mmHg)
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002297
CAS Number: 13020-57-0
EINECS: 235-879-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty