Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Methionine sulfoxide 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Amino-4-(methylsulfinyl)butanoic acid2-Amino-4-(methylsulphinyl)butanoic acid

A18081-14 A18081-06 A18081-06 A18081-14
AAA18081-06EA 71.39 USD InStock
AAA18081-06 CAAAA18081-14 CAAAA18081-06 AAA18081-14
DL-Methionine sulfoxide 98+%
DL-Methionine sulphoxide


Formula: C₅H₁₁NO₃S
MW: 165.21 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002620
CAS Number: 62697-73-8
EINECS: 263-700-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty