Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-1H-pyrazole 97+%

Supplier: Thermo Scientific Chemicals
Danger

A18083-03 A18083-06 A18083-03 A18083-06
AAA18083-06EA 324.99 USD InStock
AAA18083-06 CAAAA18083-03 CAAAA18083-06 AAA18083-03
4-Methyl-1H-pyrazole 97+%
4-Methyl-1H-pyrazole


Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 204…207 °C
Melting Pt: 13 °C
Density: 1.018
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005245
CAS Number: 7554-65-6
EINECS: 231-445-0
UN: 1760
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty