Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propyl formate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Formic acid propyl ester

A18090-AP A18090-AP
AAA18090-APEA 129.99 USD InStock
CAAAA18090-AP AAA18090-AP
Propyl formate 97%
Propyl formate


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 80…81 °C
Melting Pt: –93 °C
Density: 0.904
Flash Pt: -4 °C(25 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003295
CAS Number: 110-74-7
EINECS: 203-798-0
UN: 1281
ADR: "3",II
Merck Index: 12,08043
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty