Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cacodylic acid sodium salt trihydrate ≥98%, ee 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dimethylarsonic acid sodium salt trihydrateSodium cacodylate trihydrateSodium dimethylarsinate trihydrateDimethylarsinic acid sodium salt trihydrate

A18139-36 A18139-14 A18139-36 A18139-14 A18139-22 A18139-22
AAA18139-36EA 924.99 USD InStock
AAA18139-36 AAA18139-14 CAAAA18139-14 CAAAA18139-36 CAAAA18139-22 AAA18139-22
Cacodylic acid sodium salt trihydrate ≥98%, ee 98%
Cacodylic acid sodium salt trihydrate
An integral component of terminal deoxynucleotidyl transferase buffer; also useful in electron microscopy as a bactericide buffer


Formula: (CH₃)₂As(O)ONa·3H₂O
MW: 213.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149079
CAS Number: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty