Synonyms:
Dimethylarsonic acid sodium salt trihydrate
, Sodium cacodylate trihydrate
, Sodium dimethylarsinate trihydrate
, Dimethylarsinic acid sodium salt trihydrate
A18139-36
A18139-14
A18139-36
A18139-14
A18139-22
A18139-22
AAA18139-36EA
924.99
USD
InStock
AAA18139-36
AAA18139-14
CAAAA18139-14
CAAAA18139-36
CAAAA18139-22
AAA18139-22
Cacodylic acid sodium salt trihydrate ≥98%, ee 98%
Cacodylic acid sodium salt trihydrate
An integral component of terminal deoxynucleotidyl transferase buffer; also useful in electron microscopy as a bactericide buffer