Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

A18151-14 A18151-06 A18151-06 A18151-14
AAA18151-06EA 81.19 USD InStock
CAAAA18151-06 AAA18151-06 AAA18151-14 CAAAA18151-14
2-Bromobenzylamine hydrochloride 97%
2-Bromobenzylamine hydrochloride


Formula: C₇H₉BrClN
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012853
CAS Number: 5465-63-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty