Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: Bz-Arg-OEt.HCl

A18181-06 A18181-06 A18181-03 A18181-03
AAA18181-06EA 67.09 USD InStock
AAA18181-06 AAA18181-03 CAAAA18181-03 CAAAA18181-06
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride


Formula: C₁₅H₂₂N₄O₃
MW: 342.82 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Freezer
MDL Number: MFCD00012579
CAS Number: 2645-08-1
EINECS: 220-157-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty