Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(2-Cyanoethyl)glycine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N-(2-Cyanoethyl)aminoacetic Acid

A18199-06 A18199-14 A18199-14 A18199-06
AAA18199-06EA 33.69 USD InStock
CAAAA18199-06 AAA18199-06 CAAAA18199-14 AAA18199-14
N-(2-Cyanoethyl)glycine 98%
N-(2-Cyanoethyl)glycine


Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
Melting Pt: ca 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066903
CAS Number: 3088-42-4
EINECS: 221-418-1
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty