Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4-Methoxybenzoyl)propionic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-(4-Methoxyphenyl)-4-oxobutanoic acid

A18210-14 A18210-14 A18210-22 A18210-22 A18210-06 A18210-06
AAA18210-06EA 48.28 USD InStock
CAAAA18210-06 AAA18210-06 CAAAAA18210-22 CAAAA18210-14 AAAA18210-22 AAA18210-14
3-(4-Methoxybenzoyl)propionic acid 98+%
3-(4-Methoxybenzoyl)propionic acid


Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00002795
CAS Number: 3153-44-4
EINECS: 221-593-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty