Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Carbobenzoxy-L-glutamine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Cbz-L-glutamineZ-Gln-OH

A18240-14 A18240-14 A18240-22 A18240-06 A18240-0B A18240-06
AAA18240-22EA 162.99 USD InStock
AAA18240-22 CAAAA18240-14 CAAAA18240-06 AAA18240-14 AAA18240-0B AAA18240-06
N-Carbobenzoxy-L-glutamine 99%
N-Carbobenzoxy-L-glutamine


Formula: C₁₃H₁₆N₂O₅
MW: 280.28 g/mol
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00008043
CAS Number: 2650-64-8
EINECS: 220-173-8
Order Now
Specification Test Results
Optical rotation -7° (c=2 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty