Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-1-iodobenzene

A18255-14 A18255-09 A18255-09 A18255-22 A18255-14 A18255-22
AAA18255-22EA 246.99 USD InStock
AAA18255-22 CAAAA18255-14 AAAA18255-09 AAA18255-14 CAAAAA18255-09 CAAAA18255-22
1-Chloro-2-iodobenzene 99%
1-Chloro-2-iodobenzene


Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 108…110 °C/15 mm
Melting Pt: 1 °C
Density: 1,954
Flash Pt: 111 °C(231 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001033
CAS Number: 615-41-8
EINECS: 210-423-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty