Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Heptanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-2-Hydroxy heptane (±)-Pentyl methyl carbinol (±)-Methyl pentyl carbinol

A18391-18 A18391-18 A18391-30 A18391-30
AAA18391-30EA 151.99 USD InStock
CAAAA18391-18 AAA18391-30 CAAAA18391-30 AAA18391-18
(±)-2-Heptanol 98%
(±)-2-Heptanol


Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 160…162 °C
Density: 0.819
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004587
CAS Number: 543-49-7
EINECS: 208-844-3
UN: 1987
ADR: "3",III
Merck Index: 13,04682
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty