Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inulin

Supplier: Thermo Scientific Chemicals
Synonyms: Inulin (by Enzymatic synthesis)DahlinAlantin

A18425-18 A18425-09 A18425-30 A18425-18 A18425-09 A18425-30
AAA18425-09EA 38.49 USD InStock
CAAAA18425-09 AAA18425-09 CAAAA18425-30 AAA18425-18 AAA18425-30 CAAAA18425-18
Inulin
Inulin
Mixture of fructose polymers that serves as carbohydrate storage in plants


Storage Temperature: Ambient
MDL Number: MFCD00131407
CAS Number: 9005-80-5
EINECS: 232-684-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty