Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Di-tert-butylphenol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,3-Di(dimethylethyl)-4-hydroxybenzene

A18509-30 A18509-30 A18509-0B A18509-0B
AAA18509-0BEA 76.79 USD InStock
AAA18509-0B CAAAA18509-30 CAAAA18509-0B AAA18509-30
2,4-Di-tert-butylphenol 97%
2,4-Di-tert-butylphenol


Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 265 °C
Melting Pt: 53…57 °C
Density: 0.887
Flash Pt: 129 °C(264 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008828
CAS Number: 96-76-4
EINECS: 202-532-0
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty