Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) formate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A18569-30 A18569-30 A18569-18 A18569-18
AAA18569-18EA 50.73 USD InStock
CAAAA18569-18 AAA18569-18 AAA18569-30 CAAAA18569-30
Copper(II) formate tetrahydrate 98%
Copper (II) formate tetrahydrate


Formula: C₂H₂CuO₄·4H₂O
MW: 225.65 g/mol (153.59 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00167128
CAS Number: 5893-61-8
EINECS: 208-865-8
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty