Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isophytol 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,7,11,15-Tetramethylhexadec-1-en-3-ol

A18583-09 A18583-18 A18583-18 A18583-09
AAA18583-09EA 42.59 USD InStock
CAAAA18583-09 AAA18583-09 AAA18583-18 CAAAA18583-18
Isophytol 95%
Isophytol


Formula: C₂₀H₄₀O
MW: 296.54 g/mol
Boiling Pt: 125…128 °C/0.06 mm
Density: 0.8519
Storage Temperature: Ambient
MDL Number: MFCD00048380
CAS Number: 505-32-8
EINECS: 208-008-8
Merck Index: 13,05217
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty